siRNA Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191809.1 | NP_001178738.1 | 9 | 2148 | ||
XM_006245023.2 | XP_006245085.1 | 12 | 2517 | |||
XM_006245024.2 | XP_006245086.1 | 11 | 3850 | |||
XM_006245025.2 | XP_006245087.1 | 11 | 2413 | |||
XM_006245026.2 | XP_006245088.1 | 12 | 2760 | |||
XM_006245029.2 | XP_006245091.1 | 12 | 2627 | |||
XM_006245031.2 | XP_006245093.1 | 11 | 2523 | |||
XM_006245032.2 | XP_006245094.1 | 11 | 2656 | |||
XM_006245033.2 | XP_006245095.1 | 11 | 2432 | |||
XM_006245034.2 | XP_006245096.1 | 11 | 2322 |
