siRNA Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191809.1 | NP_001178738.1 | 9 | 2138 | ||
XM_006245023.2 | XP_006245085.1 | 12 | 2507 | |||
XM_006245024.2 | XP_006245086.1 | 11 | 3840 | |||
XM_006245025.2 | XP_006245087.1 | 11 | 2403 | |||
XM_006245026.2 | XP_006245088.1 | 12 | 2750 | |||
XM_006245029.2 | XP_006245091.1 | 12 | 2617 | |||
XM_006245031.2 | XP_006245093.1 | 11 | 2513 | |||
XM_006245032.2 | XP_006245094.1 | 11 | 2646 | |||
XM_006245033.2 | XP_006245095.1 | 11 | 2422 | |||
XM_006245034.2 | XP_006245096.1 | 11 | 2312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
