siRNA Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191809.1 | NP_001178738.1 | 8 | 1825 | ||
XM_006245023.2 | XP_006245085.1 | 11 | 2194 | |||
XM_006245024.2 | XP_006245086.1 | 10 | 3527 | |||
XM_006245025.2 | XP_006245087.1 | 10 | 2090 | |||
XM_006245026.2 | XP_006245088.1 | 11 | 2437 | |||
XM_006245029.2 | XP_006245091.1 | 11 | 2304 | |||
XM_006245031.2 | XP_006245093.1 | 10 | 2200 | |||
XM_006245032.2 | XP_006245094.1 | 10 | 2333 | |||
XM_006245033.2 | XP_006245095.1 | 10 | 2109 | |||
XM_006245034.2 | XP_006245096.1 | 10 | 1999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
