siRNA Details
Gene Symbol: Smarcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108222.1 | NP_001101692.1 | 9 | 1752 | ||
XM_006245212.2 | XP_006245274.1 | 10 | 2164 | |||
XM_006245213.2 | XP_006245275.1 | 10 | 1688 | |||
XM_008767235.1 | XP_008765457.1 | 10 | 1828 | |||
XM_008767236.1 | XP_008765458.1 | 10 | 2052 | |||
XM_008767237.1 | XP_008765459.1 | 10 | 1198 | |||
GenBank mRNA | BC168154.1 | - | 10 | 1766 | 8362162 |
