siRNA Details
Gene Symbol: Smarcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108222.1 | NP_001101692.1 | 13 | 2315 | ||
XM_006245212.2 | XP_006245274.1 | 14 | 2727 | |||
XM_006245213.2 | XP_006245275.1 | 14 | 2251 | |||
XM_008767235.1 | XP_008765457.1 | 14 | 2391 | |||
XM_008767236.1 | XP_008765458.1 | 14 | 2615 | |||
XM_008767237.1 | XP_008765459.1 | 14 | 1761 | |||
GenBank mRNA | BC168154.1 | - | 14 | 2329 | 8362162 |
