siRNA Details
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199112.1 | NP_954543.1 | 3, 4 | 500 | ||
XM_006245226.2 | XP_006245288.1 | 3, 4 | 534 | |||
GenBank mRNA | BC061750.1 | - | 3, 4 | 500 | 5599704 |

More Information
Other Species Targeted

Gene Ontology Categories:
