siRNA Details
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024891.1 | NP_001020062.1 | 6, 7 | 1118 | ||
XM_008767250.1 | XP_008765472.1 | 6, 7 | 1758 | |||
GenBank mRNA | BC097416.1 | - | 6, 7 | 1118 | 7454359 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001122685, NM_029777, AK076751, BC005518, AK029751, and others.
Gene Ontology Categories:
