siRNA Details
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024891.1 | NP_001020062.1 | 6 | 990 | ||
XM_008767250.1 | XP_008765472.1 | 6 | 1630 | |||
GenBank mRNA | BC097416.1 | - | 6 | 990 | 7454359 |
