siRNA Details
Gene Symbol: Trip12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031659.1 | NP_001026829.1 | 17 | 2572 | ||
XM_006245377.2 | XP_006245439.1 | 18 | 2681 | |||
XM_006245379.2 | XP_006245441.1 | 18 | 2678 | |||
XM_006245380.2 | XP_006245442.1 | 18 | 2681 | |||
XM_006245386.1 | XP_006245448.1 | 17 | 2597 | |||
XM_008767252.1 | XP_008765474.1 | 18 | 2681 | |||
XM_008767253.1 | XP_008765475.1 | 18 | 2732 | |||
XM_008767254.1 | XP_008765476.1 | 18 | 2678 | |||
XM_008767255.1 | XP_008765477.1 | 18 | 2681 | |||
XM_008767256.1 | XP_008765478.1 | 19 | 2594 | |||
XM_008767257.1 | XP_008765479.1 | 17 | 2600 | |||
GenBank mRNA | BC100624.1 | - | 17 | 2572 | 7930421 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133975, AK158807, AK041991.
Gene Ontology Categories:
