siRNA Details
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108230.1 | NP_001101700.1 | 13 | 1514 | ||
XM_006245396.2 | XP_006245458.2 | 17 | 5919 | |||
XM_006245397.2 | XP_006245459.2 | 17 | 5883 | |||
XM_006245398.2 | XP_006245460.2 | 16 | 5804 | |||
XM_006245400.2 | XP_006245462.2 | 15 | 5771 | |||
XM_006245401.2 | XP_006245463.2 | 16 | 5760 | |||
XM_006245402.2 | XP_006245464.2 | 16 | 5703 | |||
XM_006245403.2 | XP_006245465.2 | 14 | 5612 |
