siRNA Details
Gene Symbol: Mlph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012135.1 | NP_001012135.1 | 7 | 992 | ||
XM_006245404.2 | XP_006245466.1 | 7 | 1193 | |||
XM_006245405.2 | XP_006245467.1 | 7 | 1195 | |||
GenBank mRNA | BC081894.1 | - | 7 | 992 | 7110013 |
