siRNA Details
Gene Symbol: Ndufa10l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182671.2 | NP_872612.1 | 1 | 158 | ||
GenBank mRNA | AY260163.1 | - | 1 | 158 |
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199495.2 | NP_955789.2 | 2 | 196 | ||
XM_008767385.1 | XP_008765607.1 | 2 | 208 | |||
GenBank mRNA | BC093375.1 | - | 2 | 155 | 7389276 | |
CB610197.1 | - | 2 | 196 | |||
FQ209525.1 | - | 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
