siRNA Details
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108239.1 | NP_001101709.1 | 17, 18 | 2585 | ||
XM_008767114.1 | XP_008765336.1 | 16, 17 | 2614 |

More Information
Other Species Targeted

Gene Ontology Categories:
