siRNA Details
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108240.1 | NP_001101710.1 | 7 | 1036 | ||
XM_006227791.2 | XP_006227853.1 | 7 | 1038 |

More Information
Other Species Targeted

Gene Ontology Categories:
