siRNA Details
Gene Symbol: RGD1309621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237707.2 | XP_006237769.1 | 3 | 2690 | ||
XM_006237708.2 | XP_006237770.1 | 3 | 2674 | |||
XM_008763457.1 | XP_008761679.1 | 3 | 2691 |
