siRNA Details
Gene Symbol: Pcdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169129.1 | NP_001162600.1 | 3 | 2522 | ||
XM_006257222.2 | XP_006257284.1 | 4 | 2863 | |||
XM_006257223.2 | XP_006257285.1 | 4 | 2863 | |||
XM_006257224.2 | XP_006257286.1 | 3 | 2722 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
