siRNA Details
Gene Symbol: Apool | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014105.1 | NP_001014127.1 | 6 | 550 | ||
XM_006257191.2 | XP_006257253.1 | 6 | 489 | |||
GenBank mRNA | BC086434.1 | - | 6 | 550 | 7302620 | |
FQ219134.1 | - | 6 | 440 | |||
FQ235331.1 | - | 6 | 436 |
