siRNA Details
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191721.1 | NP_001178650.1 | 3 | 406 | ||
XM_008773343.1 | XP_008771565.1 | 3 | 424 | |||
XM_008773344.1 | XP_008771566.1 | 3 | 424 |
