siRNA Details
Gene Symbol: Uprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257154.2 | XP_006257216.1 | 6 | 896 | ||
XM_228538.8 | XP_228538.3 | 7 | 848 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Uprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257154.2 | XP_006257216.1 | 6 | 896 | ||
XM_228538.8 | XP_228538.3 | 7 | 848 |