siRNA Details
Gene Symbol: Nap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024789.1 | NP_001019960.1 | 1 | 559 | ||
GenBank mRNA | BC094525.1 | - | 1 | 559 | 7318126 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024789.1 | NP_001019960.1 | 1 | 559 | ||
GenBank mRNA | BC094525.1 | - | 1 | 559 | 7318126 |