siRNA Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108245.2 | NP_001101715.1 | 3 | 197 | ||
GenBank mRNA | FM116382.1 | - | 3 | 156 | ||
FM118664.1 | - | 3 | 181 | |||
FQ216044.1 | - | 3 | 196 | |||
FQ216100.1 | - | 3 | 173 | |||
FQ216185.1 | - | 3 | 194 | |||
FQ224925.1 | - | 3 | 194 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013712, AK003906, BC030035, AF140691.
Gene Ontology Categories:
