siRNA Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108245.2 | NP_001101715.1 | 3 | 217 | ||
GenBank mRNA | FM116382.1 | - | 3 | 176 | ||
FM118664.1 | - | 3 | 201 | |||
FQ215476.1 | - | 3 | 212 | |||
FQ216044.1 | - | 3 | 216 | |||
FQ216100.1 | - | 3 | 193 | |||
FQ224925.1 | - | 3 | 214 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013712, AK003906, BC030035, AF140691.
Gene Ontology Categories:
