siRNA Details
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014107.1 | NP_001014129.1 | 4 | 330 | ||
XM_006257631.2 | XP_006257693.1 | 3 | 376 | |||
XM_006257632.2 | XP_006257694.1 | 4 | 415 | |||
XM_006257633.2 | XP_006257695.1 | 3 | 373 | |||
XM_006257634.2 | XP_006257696.1 | 3 | 373 | |||
GenBank mRNA | BC086521.1 | - | 4 | 330 | 7302680 |
Gene Symbol: LOC686087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251400.2 | XP_006251462.1 | 1 | 348 | ||
XM_008770405.1 | XP_008768627.1 | 1 | 348 |

More Information
Other Species Targeted

Gene Ontology Categories:
