siRNA Details
Gene Symbol: Plac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024894.1 | NP_001020065.1 | 556 | |||
GenBank mRNA | BC097979.1 | - | 556 | 7443160 | ||
FQ219898.1 | - | 1 | 248 | |||
FQ228726.1 | - | 849 |
Gene Symbol: LOC102550080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257666.2 | XP_006257728.1 | 568 | |||
XM_006257669.2 | XP_006257731.1 | 3 | 550 |
