siRNA Details
Gene Symbol: MGC114492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024895.1 | NP_001020066.1 | 4 | 561 | ||
XM_008773566.1 | XP_008771788.1 | 2 | 546 | |||
GenBank mRNA | BC097440.1 | - | 4 | 561 | 7455831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
