siRNA Details
Gene Symbol: Porcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173355.1 | NP_001166826.1 | 10 | 1055 | ||
XM_006256716.2 | XP_006256778.1 | 9 | 1079 | |||
XM_006256717.2 | XP_006256779.1 | 9 | 1076 | |||
GenBank mRNA | BC158642.1 | - | 6 | 593 | 8362532 |
