siRNA Details
Gene Symbol: Ccnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773097.1 | XP_008771319.1 | 6 | 1966 | ||
XM_008773098.1 | XP_008771320.1 | 6 | 1962 | |||
XM_008773099.1 | XP_008771321.1 | 6 | 1966 | |||
XM_228779.6 | XP_228779.4 | 6 | 1966 | |||
GenBank mRNA | HQ634292.1 | - | 5 | 1849 |
