siRNA Details
Gene Symbol: Bhlhb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207611.1 | NP_997494.1 | 1 | 1624 | ||
XM_006257273.2 | XP_006257335.1 | 4 | 4430 | |||
XM_006257274.2 | XP_006257336.1 | 5 | 4281 | |||
XM_006257275.2 | XP_006257337.1 | 4 | 3244 | |||
XM_006257276.2 | XP_006257338.1 | 5 | 3392 | |||
XM_008773421.1 | XP_008771643.1 | 4 | 3226 | |||
GenBank mRNA | AF497483.1 | - | 1 | 1624 |
