siRNA Details
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108254.1 | NP_001101724.1 | 10 | 1634 | ||
XM_008773171.1 | XP_008771393.1 | 11 | 1779 | |||
XM_008773172.1 | XP_008771394.1 | 11 | 1776 | |||
GenBank mRNA | BC081944.1 | - | 11 | 1705 |
