siRNA Details
Gene Symbol: Apex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079892.1 | NP_001073361.1 | 6 | 1232 | ||
GenBank mRNA | BC127530.1 | - | 6 | 1232 | 7126449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
