siRNA Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173840.1 | NP_776212.1 | 6 | 302 | ||
XM_006249160.2 | XP_006249222.1 | 7 | 473 | |||
XM_008769138.1 | XP_008767360.1 | 6 | 755 | |||
GenBank mRNA | AY170849.1 | - | 6 | 302 | ||
FQ231408.1 | - | 4 | 162 | |||
FQ235223.1 | - | 4 | 162 |

More Information
Other Species Targeted

Gene Ontology Categories:
