siRNA Details
Gene Symbol: Yrdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175604.2 | NP_783194.2 | 4 | 753 | ||
GenBank mRNA | BC088428.1 | - | 4 | 718 | ||
BC099797.1 | - | 4 | 753 | 7936110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
