siRNA Details
Gene Symbol: Myod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176079.1 | NP_788268.1 | 1 | 309 | ||
GenBank mRNA | BC127480.1 | - | 1 | 289 | 8366406 |

More Information
Other Species Targeted

Gene Ontology Categories:
