siRNA Details
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178096.3 | NP_835197.1 | 3 | 167 | ||
XM_006254595.1 | XP_006254657.1 | 4 | 266 | |||
GenBank mRNA | AB072344.1 | - | 3 | 172 | ||
AY724475.1 | - | 4 | 198 | |||
FQ214488.1 | - | 4 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
