siRNA Details
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178096.3 | NP_835197.1 | 2, 3 | 151 | ||
XM_006254595.1 | XP_006254657.1 | 3, 4 | 250 | |||
GenBank mRNA | AB072344.1 | - | 2, 3 | 156 | ||
AY724475.1 | - | 3, 4 | 182 | |||
FQ214488.1 | - | 3, 4 | 311 |
