siRNA Details
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181388.2 | NP_852053.1 | 11, 12 | 1612 | ||
XM_008772619.1 | XP_008770841.1 | 12, 13 | 1730 | |||
XM_008772620.1 | XP_008770842.1 | 12, 13 | 1704 | |||
XM_008772621.1 | XP_008770843.1 | 11, 12 | 1474 | |||
GenBank mRNA | AY278739.1 | - | 11, 12 | 1612 | ||
FQ224821.1 | - | 12, 13 | 1735 |
