siRNA Details
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182675.1 | NP_872616.1 | 1, 2 | 145 | ||
XM_006245981.2 | XP_006246043.2 | 2, 3 | 242 | |||
GenBank mRNA | AY190240.1 | - | 1, 2 | 145 | ||
BC128720.1 | - | 1, 2 | 186 | 8374477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
