siRNA Details
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 5 | 481 | ||
XM_008765754.1 | XP_008763976.1 | 5 | 499 | |||
XM_008765755.1 | XP_008763977.1 | 5 | 499 | |||
GenBank mRNA | AY327411.1 | - | 5 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
