siRNA Details
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182844.1 | NP_878264.1 | 9 | 1481 | ||
XM_006244110.2 | XP_006244172.1 | 9 | 1521 | |||
XM_008766684.1 | XP_008764906.1 | 10 | 1599 | |||
XM_008766685.1 | XP_008764907.1 | 11 | 1713 | |||
XM_008766686.1 | XP_008764908.1 | 9 | 1523 | |||
XM_008766687.1 | XP_008764909.1 | 8 | 1994 | |||
GenBank mRNA | AY331984.1 | - | 9 | 1481 |
