siRNA Details
Gene Symbol: Wfdc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003706.3 | NP_001003706.2 | 3 | 258 | ||
GenBank mRNA | AB101676.1 | - | 3 | 274 | ||
BG661061.1 | - | 3 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
