siRNA Details
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022183.2 | NP_071519.2 | 8 | 991 | ||
GenBank mRNA | Z19552.1 | - | 8 | 991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
