siRNA Details
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029902.2 | NP_001025073.2 | 7 | 1105 | ||
XM_008758616.1 | XP_008756838.1 | 7 | 1302 | |||
XM_008758617.1 | XP_008756839.1 | 7 | 1301 | |||
XM_008758618.1 | XP_008756840.1 | 7 | 1301 | |||
XM_008758619.1 | XP_008756841.1 | 7 | 1309 | |||
XM_008758620.1 | XP_008756842.1 | 7 | 1309 | |||
GenBank mRNA | AB297790.1 | - | 7 | 1105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
