siRNA Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033898.1 | NP_001029070.1 | 2, 3 | 272 | ||
GenBank mRNA | AJ001929.1 | - | 2, 3 | 305 | ||
BC101908.1 | - | 2, 3 | 272 | 7933832 | ||
FQ222101.1 | - | 1, 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
