siRNA Details
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133543.3 | NP_598227.3 | 838 | |||
GenBank mRNA | BC088090.1 | - | 838 | 7368214 | ||
BC098650.1 | - | 900 | 7371059 | |||
FQ209576.1 | - | 822 | ||||
FQ209710.1 | - | 857 | ||||
U18762.1 | - | 1047 | ||||
U33501.1 | - | 819 |
