siRNA Details
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005560.1 | NP_001005560.1 | 9 | 1424 | ||
XM_006241997.2 | XP_006242059.1 | 9 | 2246 | |||
XM_006241998.2 | XP_006242060.1 | 9 | 2200 | |||
XM_006241999.2 | XP_006242061.1 | 9 | 2246 | |||
XM_006242000.2 | XP_006242062.1 | 9 | 1925 | |||
XM_006242001.2 | XP_006242063.1 | 10 | 1557 | |||
XM_006242002.1 | XP_006242064.1 | 9 | 1793 | |||
XM_006242003.1 | XP_006242065.1 | 10 | 1865 | |||
XM_006242004.2 | XP_006242066.1 | 9 | 1486 | |||
XM_008765756.1 | XP_008763978.1 | 10 | 1833 | |||
GenBank mRNA | BC081916.1 | - | 9 | 1424 | 7112407 |
