siRNA Details
Gene Symbol: Fopnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108261.1 | NP_001101731.1 | 2 | 204 | ||
XM_006245848.2 | XP_006245910.1 | 2 | 233 | |||
GenBank mRNA | BC083733.1 | - | 2 | 178 | ||
BC158662.1 | - | 2 | 170 | 8369763 | ||
FQ215640.1 | - | 2 | 170 | |||
FQ228774.1 | - | 2 | 178 | |||
FQ228837.1 | - | 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
