siRNA Details
Gene Symbol: LOC100362572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767466.1 | XP_008765688.1 | 1, 2 | 381 | ||
XM_008767467.1 | XP_008765689.1 | 1, 2 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
