siRNA Details
Gene Symbol: LOC100362572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767466.1 | XP_008765688.1 | 4 | 582 | ||
XM_008767467.1 | XP_008765689.1 | 3 | 567 | |||
XM_008767468.1 | XP_008765690.1 | 4 | 317 | |||
XM_008767469.1 | XP_008765691.1 | 4 | 259 | |||
XM_008767470.1 | XP_008765692.1 | 4 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
