siRNA Details
Gene Symbol: LOC360479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014115.1 | NP_001014137.1 | 4 | 571 | ||
XM_006245807.2 | XP_006245869.1 | 4 | 791 | |||
XM_006245809.2 | XP_006245871.1 | 4 | 624 | |||
XM_006245810.2 | XP_006245872.1 | 4 | 770 | |||
GenBank mRNA | BC083674.1 | - | 4 | 571 | 7135711 |
