siRNA Details
Gene Symbol: Dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038595.1 | NP_001033684.1 | 4, 5 | 627 | ||
NM_001038596.1 | NP_001033685.1 | 4, 5 | 627 | |||
XM_006245817.2 | XP_006245879.1 | 4, 5 | 667 | |||
GenBank mRNA | AF516338.1 | - | 4, 5 | 627 | ||
AY077460.1 | - | 4, 5 | 627 |
